This item is now discontinued. Please browse similar items
KNF K860-0XTK Service Kit for LABOPORT® Diaphragm Vacuum Pumps 79164-53, -55, -56, and -57
Representative image only.
KNF Mfr # K860-0XTK – Item # EW-78162-94
Service to keep your vacuum running at its best!
SPECS & DESCRIPTION
Specifications & Description
More About this Item
These service kits help keep your diaphragm vacuum pumps running smoothly. The kits only service one pump head. Two service kits are required for two-stage pumps.