KNF N035 STI-EX Explosion-proof vacuum pump, 1.2 cfm, 27" Hg, 115 VAC

Representative image only.
KNF Mfr # N035 STI-EX – Item # EW-07056-50
Oversized, permanently lubricated bearings for long-term operation
  • Ideal for hazardous areas where flammable gases or vapors may be present.
  • Pumps 07056-50 and 78164-16 are rated for uses in Class I, Group D, Division 1 environments.
  • Pumps 78164-10, -12, -14, and -20 are rated for use in Class I, Groups C & D, Division 1 environments.
  • 1 year warranty

$2,647.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Wetted PartsPTFE, 316 Stainless Steel
  • Free-Air Capacity (CFM)1.2
  • Free-Air Capacity (LPM)34
  • Free-Air Capacity (LPH)2040
  • Max Flow Rate (CFM)1.16
  • Max Flow Rate (Liters/min)33
  • Max Vacuum (in Hg)27
  • Max Vacuum (mbar)100
  • Max Vacuum (mm Hg)74.2
  • Max Vacuum (torr)74.2
  • Port Size1/4" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115
  • Power (Hz)60
  • Height (in)9.25
  • Length (in)13.75
  • Height (cm)23.5
  • Length (cm)34.93
  • Height (mm)235
  • Length (mm)349.3
  • DescriptionExplosion-proof vacuum pump, 1.2 cfm, 27" Hg, 115 VAC
  • Warranty1 year
1 Year Warranty

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present.
  • Pumps 07056-50 and 78164-16 are rated for uses in Class I, Group D, Division 1 environments.
  • Pumps 78164-10, -12, -14, and -20 are rated for use in Class I, Groups C & D, Division 1 environments.
  • 1 year warranty

More About this Item

See page 1258 for more information on National Electrical Code (NEC) hazardous classifications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat