Kobold S-3108 0.3-2.3 Gpm Brass 1/4 Innpt Float Type Flow Switch
Representative image only.
Kobold Mfr # S-3108 – Item # EW-32481-45
Small, compact design with low weight
  • Constructed of durable brass
$290.70USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)160
  • Max Operating Temperature (° C)71
  • Description0.3-2.3 Gpm Brass 1/4 Innpt Float Type Flow Switch
Key Features
  • Constructed of durable brass
More About this Item
The device works on the well known float principle but without the use of the open conical measuring tube. Instead, the instruments use a cylindrical control tube with a conical groove along its axis. The flow switch can be used for continuous control of fluids and gases to ensure that excess or low flow cannot result in damage to the equipment. Applications can be found wherever it is necessary to control flow reliably.
FEATURES
  • ± 4% Full scale accuracy
  • Small, compact design, low weight
  • Direct reading scales for water and air
  • Large and small measuring ranges from 28:1 to 2.6:1 turndown
  • Vertical connections for easy installation
  • All media contacted parts available in brass or stainless steel construction