Kodiak 133299 Liquid Level Switch Ls-1900 All PTFE 0/300F 40PSIG Spdt 20VA
Representative image only.
Kodiak Mfr # 133299 – Item # EW-68964-25
$528.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSI)40
  • DescriptionLiquid Level Switch Ls-1900 All PTFE 0/300F 40PSIG Spdt 20VA
More About this Item
All PTFE switches operate dependably in sticky or contaminated liquids. The problem of material build-up, which inhibits float travel, is minimized or eliminated. Compatible with a broad range of chemicals. Ideal for chemical metering pumps or processing and in plating systems.
FEATURES
  • Ideal for use in viscous or contaminated liquids.
  • Liquid Specific Gravity: .90 min.
  • No metal parts to corrode
  • Mounts vertically to top or bottom of tanks
  • 1-year warranty