Kodiak 26790 Compact Level Switch, Brass PTFE Float 20VA Normally Open
Representative image only.
Kodiak Mfr # 26791 – Item # EW-68964-21
$119.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCompact Level Switch, Brass PTFE Float 20VA Normally Open
More About this Item
High or Low Liquid Level
Extremely compact, small float displacement, 20VA switch. Whenever possible, single station units with greater float displacement and switch ratings should be used. For use in oils, water. Supplied with 24", #22AWG lead wires.