Koehler K28100 Core-Type Sampler
Representative image only.
Koehler Mfr # K28100 – Item # EW-17301-48
Simple spring mechanism for taking accurate samples
  • Conforms to the specifications of D4057
  • Obtain bottom samples or samples from any level
  • Butterfly valve on the bottom for ease of sampling
  • Robust stainless steel and brass construction
  • Three petcocks for draining at different levels
$931.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel, brass, polycarbonate
  • DescriptionCore-Type Sampler
Key Features
  • Conforms to the specifications of D4057
  • Obtain bottom samples or samples from any level
  • Butterfly valve on the bottom for ease of sampling
  • Robust stainless steel and brass construction
  • Three petcocks for draining at different levels
More About this Item
This core-type crude oil sampler will obtain samples from nearly any level. Use to manually obtain samples of a liquid, semi-liquid, or solid state whose vapor pressure at ambient conditions is below 101 kPa.
Rugged stainless steel and brass construction ensures longevity. Durable, 16” (40 cm) clear polycarbonate sample container lets you visually determine the amount of sediment and water. The butterfly valve on the bottom allows for easy sampling, and three petcocks along the column permit efficient draining at different levels. The petcocks are located along the column at 4", 8" and 12" (10, 20, and 30 cm).
The Core-Type Sampler is used to manually obtain samples of a liquid. semi liquid or solid state whose vapor pressure or ambient conditions is below 101kPa (crude oil, etc), in accordance to ASTM D4057.