Koflo 1/2-12PPT Mounted in-line mixer; 1/2" tube OD, 12 elements, 10/pack
Representative image only.
Koflo Mfr # 1/2-12PPT – Item # EW-04668-06
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyacetal
  • No. Of Elements12
  • Tubing OD (in)0.5
  • Tubing OD (mm)12.7
  • Mixer Length (in)6.5
  • Mounting Type0.5\" plain end
  • DescriptionMounted in-line mixer; 1/2\" tube OD, 12 elements, 10/pack
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Mounted mixers can be used with any compression fitting with the same tubing OD.