Koflo 1.5-80-10-12-2 Pipe Mixer, Kynar schedule 80, 1-1/2" NPT(M), 12 elements
Representative image only.
Koflo Mfr # 1.5-80-10-12-2 – Item # EW-04669-40
Mix fluids as you pump them through a pipe line
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPVDF
  • No. Of Elements12
  • Pipe Id (in)1.5
  • Pipe Id (mm)38.1
  • Mixer Length (in)24
  • Mounting Type1.5\" NPT(M)
  • DescriptionPipe Mixer, Kynar schedule 80, 1-1/2\" NPT(M), 12 elements
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.