Koflo 1.5-SA-3-12RH-11 Sanitary Mixer, helical design, 1-1/2" Tri-clamp size, 12 elements
Representative image only.
Koflo Mfr # 1.5-SA-3-12RH-11 – Item # EW-04668-82
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • No. Of Elements12
  • Pipe Id (in)1.37
  • Pipe Id (mm)34.8
  • Mixer Length (in)27
  • Mounting Type1.5\" Tri-Clamp
  • DescriptionSanitary Mixer, helical design, 1-1/2\" Tri-clamp size, 12 elements
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Helical Design Mixers are best for high-viscosity (>1,500 Cp) fluids, and when fluids are moving more slowly through the mixer. Excellent for combining products into fluids such as yogurt, mayonnaise, and ice cream.