Koflo 1-80-10-6-2 Pipe Mixer, Kynar schedule 80, 1" NPT(M), 6 elements

Representative image only.
Koflo Mfr # 1-80-10-6-2 – Item # GY-04669-32
Mix fluids as you pump them through a pipe line

PRODUCT OPTIONS

$398.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPVDF
  • No. Of Elements6
  • Pipe Id (in)1.05
  • Pipe Id (mm)26.67
  • Mixer Length (in)8
  • Mounting Type1" NPT(M)
  • DescriptionPipe Mixer, Kynar schedule 80, 1" NPT(M), 6 elements

More About this Item

Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat