Koflo 3/16-24P Unmounted in-line mixer; 3/16"OD, 24 elements, 10/pack
Representative image only.
Koflo Mfr # 3/16-24P – Item # EW-04667-12
Works well with flexible tubing
  • Limited Lifetime Warranty
$67.50USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyacetal
  • No. Of Elements24
  • Tubing ID (in)0.1875
  • Tubing ID (mm)4.76
  • Mixer Length (in)4.625
  • DescriptionUnmounted in-line mixer; 3/16\"OD, 24 elements, 10/pack
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Unmounted mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table. They work well with flexible tubing.