Koflo 5/8-24PPT Mounted in-line mixer; 5/8" tube OD, 24 elements, 10/pack
Representative image only.
Koflo Mfr # 5/8-24PPT – Item # GY-04668-18
$126.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyacetal
  • No. Of Elements24
  • Tubing OD (in)0.625
  • Tubing OD (mm)15.88
  • Mixer Length (in)14.25
  • Mounting Type0.625" plain end
  • DescriptionMounted in-line mixer; 5/8" tube OD, 24 elements, 10/pack
More About this Item
Mounted mixers can be used with any compression fitting with the same tubing OD.