Kynar Luer Fitting, PVDF, Two-Way Stopcock, Female Luer Lock x Male Luer Lock; 1/Ea
Representative image only.
Kynar Mfr # K-LL2W – Item # EW-31200-70
Build leakproof fluid handling systems
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$52.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortFemale Luer Lock
  • Inlet PortMale Luer Lock
  • Wetted MaterialsNatural PVDF, Polypropylene
  • Flow PatternTwo-Way
  • DescriptionLuer Fitting, PVDF, Two-Way Stopcock, Female Luer Lock x Male Luer Lock
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
More About this Item
These one-way stopcocks feature an all-black PVDF construction.