L-K Industries LK-TB200 Liquid Petroleum Gas Corrosion Test Cylinder, 1/ea
Representative image only.
L-K Industries Mfr # LK-TB200 – Item # EW-17700-85
Required for use with LPG Corrosion Test Cylinder Heater
  • Conforms to ASTM D1838 and NPGA Publication 2140 standards
  • Constructed of durable stainless steel for noncorrosive product applications
  • Designed especially for conducting ASTM and NGPA copper strip corrosion tests of liquefied petroleum gas
$524.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithL-K Industries LPG Corrosion Test Cylinder Heater
  • DescriptionLiquid Petroleum Gas Corrosion Test Cylinder, 1/ea
Key Features
  • Conforms to ASTM D1838 and NPGA Publication 2140 standards
  • Constructed of durable stainless steel for noncorrosive product applications
  • Designed especially for conducting ASTM and NGPA copper strip corrosion tests of liquefied petroleum gas
More About this Item
Compatible with the L-K Industries LP Gas Corrosion Test Cylinder Heater and in compliance with industry standard ASTM D1838. It is made without welds and has an O-ring seal, which will maintain pressure when tightened by hand. No wrenches or other tools are required, so the unit can be used anywhere onsite. The placement of the O-ring, a common replacement part, allows for easy access and reduces down time. It is design tested to 3000 psi.
L-K Industries LP Gas Corrosion Test Cylinder Heater are available as models 17700-40 and -41, and are sold separately.