L-K Industries Melton Oil Centrifuge for Short Cone Tubes, 220 VAC
Representative image only.
L-K Industries Mfr # D-4-220-C – Item # EW-17700-17
Versatile—each cup is individually heated and controlled
  • Separate heating elements and thermostats allow different temperature conditions at the same time
  • Built-in mechanical tachometer shows current rotational speed at a glance
  • Hand brake brings rotor to a stop
PRODUCT OPTIONS
$9,000.00USD / Each
Often Bought with...
Cole-Parmer ThinTouch™ Nitrile Gloves
$9.00 - $79.50USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Variable SpeedYes
  • Rotors IncludedFour-place
  • Temp ControlAdjustment screw
  • Tube Capacity100-mL Short cone
  • Power (VAC)220
  • Power (Hz)50
  • DescriptionOil Centrifuge for Short Cone Tubes, 220 VAC
Includes
four-place rotor.
Key Features
  • Separate heating elements and thermostats allow different temperature conditions at the same time
  • Built-in mechanical tachometer shows current rotational speed at a glance
  • Hand brake brings rotor to a stop
More About this Item
This benchtop Melton-type oil centrifuge is factory preset to keep samples heated at 150°F (66°C), but the temperature can be adjusted by simply turning an adjustment screw. Built-in mechanical tachometer and hand brake keep operation simple.
Order sample tubes and companion sample heater separately.
Note: All models must be hard-wired by a qualified electrician.