GoatThroat BGA Lab/shop air adapter for pumps
Representative image only.
GoatThroat Mfr # BGA – Item # EW-75650-40
Enhance versitility of your GoatThroat™ Spillproof Barrel Pump
PRODUCT OPTIONS
$208.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLab/shop air adapter for pumps
More About this Item
Accessories for GoatThroat Spillproof Barrel Pump optimize performance or suitable for maintenance.
The pneumatic air adapter, model 75650-40, allows GoatThroat™ pumps to be used with shop air, air compressors, or bottle compressed air. Uses up to 4 psi to deliver fluids up to 4.5 GPM. External pressure valve keeps containers from being over pressurized.