LabChem Potassium Hydroxide, 0.5 N (0.5 M) in Methanol; 1 L

Representative image only.
LabChem Mfr # LC195202 – Item # EW-80034-50
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Normality0.5
  • Purity Grade
  • Concentration0.5 M
  • ChemistrySolvents
  • MediumMethanol
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationClass 3 (8), Packing Group II
  • UN NumberUN2924
  • Shelf Life (days)540
  • Shelf Life (months)18
  • DescriptionPotassium Hydroxide, 0.5 N (0.5 M) in Methanol; 1 L

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat