Labconco 4596100 Mobile Conversion Kit for Washers; 115V, 60hz
Representative image only.
Labconco Mfr # 4596100 – Item # EW-98963-80
Optimize performance of your Labconco glassware washer
  • Adds more cleaning options to your washer
  • Adds mobilty to freestanding SteamScrubber and FlaskScrubber glassware washers
  • Allows washers to be plugged into a 20 amp electrical wall outlet
  • Connects to standard lab sink faucet
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionMobile Conversion Kit For Washers, 115V 60hz
  • Warranty1 year
1 Year Warranty
Includes
Four 2" diameter rubber casters, two aerator adapters for connection to standard 55/64"-27 male and 15/16"-27 female laboratory faucets, drain hose, installation instructions and 10 ft power cord with NEMA 5/20 plug for washers manufactured before October 2020.
Key Features
  • Adds more cleaning options to your washer
  • Adds mobilty to freestanding SteamScrubber and FlaskScrubber glassware washers
  • Allows washers to be plugged into a 20 amp electrical wall outlet
  • Connects to standard lab sink faucet
  • 1 year warranty
More About this Item
Choose from a wide range of rack accessories and other products to ensure top performance of your Labconco glassware washer.
Not for use with Labconco undercounter washers or FlaskScrubber Vantage Glassware Washers.