Labconco 5006300 Latex Hands for gGlove Boxes; size 9; 12/pack
Representative image only.
Labconco Mfr # 5006300 – Item # EW-09112-28
$145.00USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLatex Hands for gGlove Boxes; size 9; 12/pack
More About this Item
Order sleeves and interchangeable hands for greater versatility. Sleeves are neoprene and include O-rings and a spring. Sleeves fit 8 to 8-3/4" openings. Order sleeves and hands separately.