Labconco 7486400 170 mL Borosilicate Glass Tube PTFE-Coated Aluminum Block
Representative image only.
Labconco Mfr # 7486400 – Item # EW-13041-20
Enhance the performance of your Labconco CentriVap® Vacuum Concentrator with these accessories
  • Choose replacement parts or optional accessories based on your application needs
  • PTFE coating provides protection against harsh applications
  • Accommodates up to 8 tubes
PRODUCT OPTIONS
$2,150.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)125
  • Capacity (Gal)8-170 mL borosilicate glass tube
  • MaterialPTFE-coated aluminum
  • Compatible WithCentriVap® Centrifugal Vacuum Concentrator systems
  • Description170 mL Borosilicate Glass Tube PTFE-Coated Aluminum Block
Key Features
  • Choose replacement parts or optional accessories based on your application needs
  • PTFE coating provides protection against harsh applications
  • Accommodates up to 8 tubes
More About this Item
Choose from a wide range of rotors and other accessories suitable for use with your Labconco CentriVap® Complete™ Vacuum Concentrator.