Labconco Canopy Hood, 72"
Representative image only.
Labconco Mfr # 7099000 – Item # EW-09072-64
Vent non-toxic materials to keep your lab safe
  • Designed to be installed on a wall or suspended from the ceiling
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Interior Width (in)72
  • Power (VAC)115
  • DescriptionCanopy Hood, 72\"
  • Warranty1 Year
1 Year Warranty
Key Features
  • Designed to be installed on a wall or suspended from the ceiling
  • 1 year warranty
More About this Item
Canopy Hoods are specifically designed to vent non-toxic materials such as heat, steam and odors from large or bulky apparatus that do not require a physical barrier such as ovens, steam baths and autoclaves. The hood may be installed on a wall or suspended from the ceiling for peninsular and corner locations. Coupled with a blower and ductwork (sold separately), the Canopy Hood can operate at a wide range of exhaust rates.