Labconco Elbow, 90°, 6" Diameter, PVC
Representative image only.
Labconco Mfr # 4708700 – Item # EW-09073-74
Designed for quick installation and minimum pressure losses
  • PVC elbows are compatible with thermoplastic duct
  • Elbows feature belled end connections to receive PVC duct directly
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Ductwork Diameter (in)6
  • DescriptionElbow, 90°, 6\" Diameter, PVC
  • Warranty1 year
1 Year Warranty
Key Features
  • PVC elbows are compatible with thermoplastic duct
  • Elbows feature belled end connections to receive PVC duct directly
  • 1 year warranty
More About this Item
These ductwork elbows provide fume hoods and other ventilated enclosures that require exhausting with ductwork elbows to optimize ventilation.