Labconco FreeZone Freeze Drying Chamber, 16 Ports, SS/PTFE-Coated Interior
Representative image only.
Labconco Mfr # 7522901 – Item # EW-03353-43
Easily attach flasks to your freeze dry system or contain samples in bulk
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Designed with a corrosion-resistant PTFE-coated interior
  • Accommodates ½” and ¾” adapters
  • Compatible with all FreeZone Freeze Dryers
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Exterior Height (InchShort)13
  • DescriptionFreeze Drying Chamber, 16 Ports, SS/PTFE-coated interior
  • Warranty1 Year
1 Year Warranty
Includes
Sixteen neoprene valves, plastic knobs, acrylic lid, neoprene gasket, and latching pin.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Designed with a corrosion-resistant PTFE-coated interior
  • Accommodates ½” and ¾” adapters
  • Compatible with all FreeZone Freeze Dryers
  • 1 year warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk. Various models are available from 8-, 12- to 16-port drying chambers to 3 to 5 heated product shelf chambers.
The stainless-steel drying chamber features 16 neoprene valves with plastic knobs that accommodate both ½” and ¾” adapters for quick and easy connection of flasks.