Labconco FreeZone Freeze Drying Chamber, 8 Ports, Clear, Tall
Representative image only.
Labconco Mfr # 7318800 – Item # EW-03353-17
Easily attach flasks to your freeze dry system or contain samples in bulk
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Obtain visibility while freeze drying samples in bulk or attach flasks
  • Accommodates tall product shelves
  • Compatible with all FreeZone Freeze Dryers
  • 1 year warranty
PRODUCT OPTIONS
$4,509.00USD / Each
$5,010.00USD / Each
$665.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Exterior Height (InchShort)23
  • DescriptionFreeze Drying Chamber, 8 Ports, Clear, Tall
  • Warranty1 Year
1 Year Warranty
Includes
Aluminum lid, 8 neoprene valves, plastic knobs, and pass through for electrical cords.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Obtain visibility while freeze drying samples in bulk or attach flasks
  • Accommodates tall product shelves
  • Compatible with all FreeZone Freeze Dryers
  • 1 year warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk. Various models are available from 8-, 12- to 16-port drying chambers to 3 to 5 heated product shelf chambers.
The tall acrylic drying chamber is ideal for freeze drying numerous samples in bulk as well as in flasks. An aluminum lid offers 8 neoprene valves with plastic knobs that accommodate both ½” and ¾” adapters for quick and easy connection of flasks. The chamber also features an 8” diameter bottom opening to the collector and fits a 10-1/8” or 14” diameter condenser.