Labconco Protector Premier 5' Fume Hood With 2 Services, No Blower; 230V/50Hz

Representative image only.
Labconco Mfr # 100500021 – Item # EW-09102-84
Excellent chemical resistance

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)60
  • Blower (hp)1/2
  • LightsFluorescent
  • Switches2
  • Exterior Width (in)60
  • Exterior Height (in)59
  • Exterior Depth (in)31.6875
  • Power (VAC)230
  • Power (Hz)50
  • Sash opening (in)37.5
  • Shipping Weight (lbs)420
  • Shipping Weight (kg)191
  • Ductwork Diameter (in)12
  • DescriptionFiberglass 60\" Fume Hood, Two Services, No Blower, 230 V, 50 Hz
Conformite EuropeenneETL Listed

More About this Item

Labconco Protector Premier Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general-purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.

These by-pass hoods feature the largest unobstructed sightline available—37.5” from work surface to header panel. Other features include Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar.

Hoods are available with a built-in standard fiberglass blower, an explosion-proof fiberglass blower, or without a blower (remote blower sold separately). Standard hoods include T8 fluorescent lighting. Explosion-proof (EP) models include incandescent lighting (bulb not included). EP blowers are UL-listed for Class I, Group D and Class II, Groups E, F and G. All models are ETL listed. 230V models are CE listed.

Additional service fixtures can be added; contact a Cole-Parmer Application Specialist.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat