Labconco Purifier 304410010 Biosafety Cabinet, 4', 230V

Representative image only.
Labconco Purifier Mfr # 304410010 – Item # EW-32521-08
Operate in recirculated filtration or exhaust mode while maintaining Class II safety
  • The world''s first Class II Biosafety Cabinet that simplifies selection
  • Improved flexibility and chemical safety over both Type A and Type B biosafety cabinets
  • Provides lifetime operational cost savings over Type B biosafety cabinets

PRODUCT OPTIONS

$18,900.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)48
  • Exterior Width (in)54.3125
  • Exterior Height (in)64
  • Exterior Depth (in)32.6875
  • Power (VAC)230
  • Power (Hz)50
  • Sash opening (in)8 and 10
  • Exhaust Volume (CFM)387 to 480
  • DescriptionBiosafety Cabinet, 4', 230V

Key Features

  • The world''s first Class II Biosafety Cabinet that simplifies selection
  • Improved flexibility and chemical safety over both Type A and Type B biosafety cabinets
  • Provides lifetime operational cost savings over Type B biosafety cabinets

More About this Item

The Purifier Axiom Class II, Type C1 is the first biological safety cabinet that can be used in a recirculating Type A-mode for standard microbiological work, or can be connected to an exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides. Because of the efficient, well-defined airflow pattern over its work surface, the Axiom is safer and easier to operate than traditional biosafety cabinets.


The Axiom provides protection for personnel, products, and the laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, with the option of venting away hazardous fumes. Type C1 enclosures may also be used for a wide variety of applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards.


The exclusive design of the Chem-Zone™ maximizes chemical safety by creating a large, clearly defined area within the enclosure, bringing peace of mind during chemical handling. Other features that make the Axiom safe and easy to operate include its ergonomic Inclination™ Technology and the intuitive MyLogic™ Operating System.


The Axiom can be run as a Type A or Type B BSC (with or without ducting). A base stand is required (not included with all models). Models available with or without options (including UV light, 1/4-turn ball valve, Vacu-Pass cord/cable port) or base stand. The 10" sash height can be adjusted to 8" for additional personal protection.

Includes efficient EC motor.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat