Labconco ScrubAir 4615700 Spindle Kit, for 7 mm Test Tubes or Vials
Representative image only.
Labconco ScrubAir Mfr # 4615700 – Item # EW-98966-06
Add versitility to your ScrubAir washer and dryer
  • Add versatility by allowing additional types of pipettes and other small glassware to be washed
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSpindle Kit, for 7 mm Test Tubes or Vials
  • Warranty1 year
1 Year WarrantyUL Listed.
Key Features
  • Add versatility by allowing additional types of pipettes and other small glassware to be washed
  • 1 year warranty
More About this Item
Spindles add versatility by permitting small test tubes or vials, 7 mm ID or larger, to be washed in the ScrubAir. Kit contains 24 each 5 mm OD polystyrene plastic spindles with caps and stainless steel test tube rack to hold up to 20 each test tubes. Compatible for use with Labconco the ScrubAir Pipette Washer/Dryer, which is sold separately.