This item is now discontinued. Please browse similar items

Labnet S2020-P4 Microplate Orbital Shaker With Timer, 115 VAC, 60 Hz

Representative image only.
Labnet Mfr # S2020-P4-B – Item # EW-51402-00
Ideal for mixing test solutions in microtitration assays
  • Securely holds four 96-well plates securely with stainless steel spring clamps

SPECS & DESCRIPTION

Specifications & Description

  • Orbit Size (in)0.125
  • Orbit Size (mm)3.2
  • Min Speed (rpm)100
  • Max Speed (rpm)1400
  • Max Load (lbs)0.66
  • Max Load (kg)0.3
  • Min Operating Temperature (° F)39
  • Max Operating Temperature (° F)149
  • Min Operating Temperature (° C)4
  • Max Operating Temperature (° C)65
  • TimerYes
  • Timer Range0 to 99 minutes
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)0.5
  • DescriptionMicroplate Orbital Shaker With Timer, 115 VAC, 60 Hz

Key Features

  • Securely holds four 96-well plates securely with stainless steel spring clamps

More About this Item

Shaker applications include hematology, urology, or EIA/ELISA immunology. Chemical resistanct contoured stainless steel platform holds plates securely.

What''''s included: a platform and a 6-1/4-ft cord with U.S. standard plug

REVIEWS

REVIEW SNAPSHOT®

by PowerReviews
Labnet S2020-P4 Microplate Orbital Shaker With Timer, 115 VAC, 60 Hz
 
5.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

 
5.0

very nice shaker

By 

from indianapolis

About Me Lab Assistant

Pros

  • Accurate
  • Easy To Use
  • Works Properly

Cons

    Best Uses

    • In The Lab
    • Research

    Comments about Labnet S2020-P4 Microplate Orbital Shaker With Timer, 115 VAC, 60 Hz:

    very nice. it could use a ruber mat for the top though

    Displaying review 1

    Back to top

    Q&A

    CUSTOMER SUPPORT

    Need Help with this product?

    Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

    Call Us At1-800-323-4340
    Live chat