Lactose broth, prepared; 500 mL bottle with 225 mL broth, 10/pk
Representative image only.
Item # EW-14204-70
Save time and money with ready-made bottles
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the cultivation of coliforms and Salmonella spp. in water, food, dairy, and pharmaceutical products
  • Single strength
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Lactose
  • Capacity (mL)225
  • Capacity (L)0.225
  • DescriptionLactose broth, prepared; 500 mL bottle with 225 mL broth, 10/pk
  • Warranty6 month
6 Month Warranty
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • For the cultivation of coliforms and Salmonella spp. in water, food, dairy, and pharmaceutical products
  • Single strength
  • 6 month warranty
More About this Item
Bottles are tested and standardized to ensure consistent and reliable performance. Each bottle includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and shelf-life guarantee. Bottle and pack sizes vary.