Large Bottle Brush 16" Radial Tip, 12/pk
Representative image only.
Item # EW-84550-81
Contoured brushes are designed to clean hard-to-reach areas
PRODUCT OPTIONS
$407.00USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester bristles, plastic handle
  • ColorWhite
  • Handle Length10
  • DescriptionLarge Bottle Brush 16" Radial Tip, 12/pk
More About this Item
Radial tip brushes allow you to remove difficult deposits without scratching your container. Folding tip and double-tufted brushes are made to fold flat through the bottle neck and expand to fill the container. Angle head brushes are manufactured to angle the brush along the curvature of the container with minimal strain. Volumetric brushes feature an angled handle which bends to fit the container shape for cleaning pear-shaped, Erlenmeyer, round bottom, flat bottom, Nephelo, Kjeldahl, filtering, and volumetric flasks.