Lascar Electronics BEZ 900-IP Waterproof Mounting Bezel for 900 Series Panel Meters
Representative image only.
Lascar Electronics Mfr # BEZ 900-IP – Item # EW-65595-05
Optimize performance of your Lascar panel meters
  • Keep your panel meters running properly with these mounting bezels and temperature probes
  • 1 year warranty
PRODUCT OPTIONS
$23.36USD / Each
$29.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithLascar 900 Series Panel Meters
  • DescriptionWaterproof Mounting Bezel for 900 Series Panel Meters
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Keep your panel meters running properly with these mounting bezels and temperature probes
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your panel meters.
When these mounting bevels are used with the Lascar 900 series panel meters, the system will have an IP67/NEMA 4X waterproof rating.