Lascar Electronics EL-USB-CASE Waterproof Protective Data Logger Case
Representative image only.
Lascar Electronics Mfr # EL-USB-CASE – Item # EW-23039-61
On-the-spot data retrieval and viewing without having to remove the logger
  • Suitable for use with EL-USB-1 and EL-USB-2
  • Rugged metal alloy design
  • Waterproof protection to IP-67/NEMA-4X
  • 1 year warranty
PRODUCT OPTIONS
$27.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialMetal Alloy
  • Compatible WithEL-USB-1 or EL-USB-2
  • DescriptionWaterproof Protective Data Logger Case
  • Warranty1 year
1 Year Warranty
Key Features
  • Suitable for use with EL-USB-1 and EL-USB-2
  • Rugged metal alloy design
  • Waterproof protection to IP-67/NEMA-4X
  • 1 year warranty
More About this Item
This is a rugged metal case that provides additional waterproof protection for the EL-USB-1 data logger, and allows the data logger to record temperatures in particularly harsh environments.
The EL-USB-2 may also benefit from the additional waterproof protection of the EL-USB-CASE, however it would be limited to measuring temperature only.