Lascar Electronics EL-WiFi-ALERT Optional Wi-Fi Alert Sensor Monitoring System
Representative image only.
Lascar Electronics Mfr # EL-WIFI-ALERT – Item # EW-23039-09
On-the-spot data retrieval and viewing without having to remove the logger
  • Detects alarms from any EL-WiFi sensor device on the same wireless network
  • Audible alarm with built-in speaker (SPL 100dBA @1m)
  • Visual alarm from high brightness LEDs
  • Continues to work when the PC is turned off
  • Easy set-up using the free PC software application
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With18000-08, 18000-09, 18000-11, 18000-12, 23039-01, 23039-02, 23039-03, 23039-07, 23039-08
  • Width (cm)3.05
  • Height (cm)9.73
  • Length (cm)7.13
  • DescriptionOptional Wi-Fi Alert Sensor Monitoring System
  • Warranty1 year
1 Year Warranty
Key Features
  • Detects alarms from any EL-WiFi sensor device on the same wireless network
  • Audible alarm with built-in speaker (SPL 100dBA @1m)
  • Visual alarm from high brightness LEDs
  • Continues to work when the PC is turned off
  • Easy set-up using the free PC software application
  • Rechargeable internal lithium polymer battery for short term backup during mains failure
  • 1 year warranty
More About this Item
The EL-WiFi-Alert is a standalone monitoring system for all sensor devices within the EasyLog WiFi range. The EL-WiFi-Alert will receive alarm messages from any EL-WiFi sensor device connected to the same wireless network.
The EL-WiFi-Alert indicates an alarm activation using high brightness LEDs and a choice of nine alarm tones. The EL-WiFi-Alert continues to work when the PC is turned off.
The EL-WiFi-Alert is designed to be continuously powered via the mains and is supplied with a universal power adaptor. However, it does contain a rechargeable lithium polymer battery for device positioning and limited power during the event of a power failure.
The WiFi signal strength indicator is activated by pressing the EL-WiFi-Alert button three times. All LEDs and a fast beep indicate a strong signal. Limited LEDs and slow beep indicate a weak signal.
The audible alarm can be muted by pressing the EL-WiFi-Alert button once or remotely from the PC software. There is also a configurable auto-mute function.
Set-up is completed through software installed on the PC. After initial set-up, audible and visual alarms can be reconfigured, wirelessly, using the PC software.
The EL-WiFi-Alert can be attached to a wall or flat surface using the bracket provided, and has a protection rating of IP44 (with cable connected).
This device is IEEE 802.11b compliant, supports WEP, WPA/WPA2 and WPA-Enterprise (*MS-CHAPv2, PEAP, EAP-FAST, EAP-TTLS).