Lascar Electronics S 43-TP Thermistor Add-On for PanelPilotACE Graphics Meter, 4.3" TFT
Representative image only.
Lascar Electronics Mfr # S 43-TP – Item # EW-18000-98
Optimize performance of your Lascar Process Control Units
  • Keep your running properly with these cables and adapters
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithSGD 43-A PanelPilotACE panel meters
  • ConnectionsFour 3.5 mm audio jacks for Thermistors (PTC or NTC), with an adjustable bias resistor value (default set at 10 kΩ)
  • DescriptionLascar Thermistor Add-On for PanelPilotACE Graphics Meter, 4.3\" TFT
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Keep your running properly with these cables and adapters
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Lascar Process Control meters