Lascar Electronics SGD ADPT-TC Thermocouple Adapter Module for PanelPilot
Representative image only.
Lascar Electronics Mfr # SGD ADPT-TC – Item # EW-18000-24
Optimize performance of your Lascar Process Control Units
  • Keep your running properly with these cables and adapters
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With18000-17, 18000-18, 18000-19
  • DescriptionThermocouple Adapter Module for PanelPilot
  • Warranty1 Year
1 Year Warranty
Key Features
  • Keep your running properly with these cables and adapters
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Lascar Process Control meters
Thermocouple add-on board comes with a beaded Type K thermocouple, but will accept any Type J, K, or T thermocouple probe.
Please see product brochure for additonal information.