Leeson 108046 NEMA Type 56C-face TENV motor, 1/3 hp, 1800 rpm, 12 VDC

Representative image only.
Leeson Mfr # 108046 – Item # EW-70072-10
Low-cost DC motors available in numerous configurations to meet your needs
  • Power pulleys, belts, pumps, or other machinery
  • Operate low-voltage models (12 or 24 VDC) with battery or solar power for fixed-speed applications
  • Explosion-proof models are Class 1, Groups C & D; Class II, Groups F & G rated

PRODUCT OPTIONS

$581.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Motor (kW)0.32
  • Max Motor RPM (rpm)1800
  • Motor Frame Size56C
  • Motor Enclosure TypeTENV
  • Power (VDC)12
  • Power Conduit Box ConnectionYes
  • Width (in)6.5
  • Length (in)11.5
  • Height (in)6.5
  • Shaft Centerline (in)3.25
  • Shaft Diameter (in)5/8
  • Width (cm)16.51
  • Length (cm)29.21
  • Height (cm)16.51
  • Shaft Centerline (cm)8.26
  • Shaft Diameter (cm)1.59
  • Motor hp1/3
  • DescriptionNEMA Type 56C-face TENV motor, 1/3 hp, 1800 rpm, 12 VDC

Key Features

  • Power pulleys, belts, pumps, or other machinery
  • Operate low-voltage models (12 or 24 VDC) with battery or solar power for fixed-speed applications
  • Explosion-proof models are Class 1, Groups C & D; Class II, Groups F & G rated

More About this Item

DC motors are suited for a variety of environments.
Totally enclosed, fan-cooled (TEFC) motors utilize an external fan to circulate outside air to the motor for applications where the motor will be running continuously.
Totally enclosed, nonvented (TENV) motors are designed without an air opening to protect against dust and other debris.
Explosion-proof (XPRF) motors are approved for areas where hazardous materials are used.
Washdown (WDN) motors are suitable for applications requiring regular washing or sanitizing cycles.
To vary the speed on a DC motor, order a DC controller at the side.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat