LeviFlow LFC-1C-PC-SK Single-Use Precision Ultrasonic Flowmeter Starter Kit
Representative image only.
LeviFlow Mfr # 100-91072 – Item # EW-24500-07
Obtain flow measurements of high-purity fluids
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 0.08 to 800 L/min
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Order starter bundle for immediate use
$2,293.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min Operating Temperature (° F)32
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° F)104
  • Max Operating Temperature (° C)40
  • Fluid TypeHigh-purity liquids
  • Viscosity0.8 to 40 cSt
  • Power24
  • Display Type4-digit LCD
  • DescriptionPrecision Ultrasonic Flowmeter Starter Kit
Key Features
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 0.08 to 800 L/min
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Order starter bundle for immediate use
More About this Item
These ultrasonic flowmeter systems are designed for use in applications were cleanliness is required. Readings are accurate to 1%—high precision for high-purity fluids. Each system requires a starter kit and clamp-on flow sensor; order separately. The starter kit includes components to get a system in place, just choose a flow sensor at the desired flow range and sterility.
Starter kit includes a single-channel flow converter with DSP for processing the sensor’s signals, connection cables for meter to PC and meter to flow sensor, power supply and cord, and configuration software on a USB stick. The converter also features a 4-digit LED display indicating the flow value.
Flow sensors feature integral sanitary Tri-Clamp® connections and are designed for one-time use. Sensors are IP65 rated and have a watertight connection box.
Order the starter bundle, which includes the starter kit and the ultrasonic sensor, for your convenience.