Lighting Specialties 3050-BK Floor Stand for Infrared Heating Lamp, with pedestal base
Representative image only.
Lighting Specialties Mfr # 3050-BK – Item # EW-03057-52
Fast, safe heat without airflow
PRODUCT OPTIONS
$305.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFloor Stand for Infrared Heating Lamp, with pedestal base
More About this Item
Mount to any table or benchtop, or use our floor stands to support the heating lamp—features durable all-metal construction. Adjust the 42" swiveling extension arm to position lamp exactly where you want it. Dual reflector prevents excessive heat buildup.
Lamp comes complete with screw-type mounting bracket; 10-ft grounded cord with plug; and 250 watt, hard glass infrared heat bulb. Easily remove wire bulb guard to replace bulb(order replacement bulb separately).
Call Us At1-847-549-7600
Live chat