Lightnin 801003 316 SS axial flow impeller ( A-100), 3.1" dia x 3/8" bore dia
Representative image only.
Lightnin Mfr # 801003 – Item # EW-04540-30
$246.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description316 SS axial flow impeller ( A-100), 3.1" dia x 3/8" bore dia
More About this Item
Axial Flow Impellers (LIGHTNIN® A-100) with SUPERPITCH® design — 1.5 pitch ratio for greater batch mixing capabilities. Standard axial flow; use for wetting dry powders.