LinkTech 50AC Quick-Disconnect Fitting Insert, w/o Valve, 1/4" Hose Barb; 25/Pk
Representative image only.
LinkTech Mfr # 50AC-PB2-04 – Item # EW-41312-13
Use with either liquid or gas—1/4" diameter flow path
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
PRODUCT OPTIONS
$21.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal (POM)
  • Valve Spring Material316 SS
  • O-Ring MaterialBuna-N
  • Number Of Ports1
  • Connection TypeQuick-Disconnect x Barb
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Quick-Disconnect Flow Size (in)1/4
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • DescriptionQuick-Disconnect Fitting Insert, Hose Barb w/o Valve, 1/4" ID; 25/pk
Key Features
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
More About this Item
The 50AC series of quick-disconnect fittings feature durable, Acetal (POM) plastic for the latch and body construction. Order bodies and inserts separately to suit your assembly needs—valved or non-valved configurations are available and compatible with each other.