LinkTech 50AC Quick-Disconnect Fitting Insert, w/o Valve, 1/4" Push-to-Connect; 25/Pk
Representative image only.
LinkTech Mfr # 50AC-PB17-04 – Item # EW-41312-39
Use with either liquid or gas—1/4" diameter flow path
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
PRODUCT OPTIONS
$120.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAcetal (POM)
  • Valve Spring Material316 SS
  • O-Ring MaterialBuna-N
  • Number Of Ports1
  • Connection TypeQuick-Disconnect x Push-to-Connect
  • Compression Size / Tube Od (Port 1) (in)1/4
  • Push-to-Connect Tube OD (port 1) (in)1/4
  • Quick-Disconnect Flow Size (in)1/4
  • Min Temperature (° F)-40
  • Max Temperature (° F)180
  • Min Temperature (° C)-40
  • Max Temperature (° C)82
  • Max Operating Pressure (PSI)120
  • DescriptionQuick-Disconnect Fitting Insert, Compression w/o Valve, 1/4" OD Push-to-Connect; 25/pk
Key Features
  • Acetal (POM) body and latch, 316 SS springs, and Buna-N O-rings for durability and compatibility
  • Mix and match valved and non-valved bodies and inserts to suit your flowpath needs
More About this Item
The 50AC series of quick-disconnect fittings feature durable, Acetal (POM) plastic for the latch and body construction. Order bodies and inserts separately to suit your assembly needs—valved or non-valved configurations are available and compatible with each other.