Logic Beach ILIM-7 Expansion Module; 8 Isolated Diff Analog In, 5VDC Out
Representative image only.
Logic Beach Mfr # ILIM-7 – Item # EW-18002-51
$1,011.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels8
  • Input TypeIsolated differential analog Inputs (IL-80)
  • DescriptionExpansion Module; 8 Isolated Diff Analog In, 5VDC Out
More About this Item
Connect the sensors, program the Internet-Logger graphically with HyperWare II, connect a communication link (Ethernet, modem, RS-232, USB, RF modem, cellular CDMA/GPRS/GSM) and you are ready to view logger-served data, system status and more from INTELLILOGGER™ provided standard web pages or custom pages specific to your application.
FEATURES
  • 8 Isolated differential analog Inputs (IL-80)
  • 3 Hi-speed analog inputs, 4 digital inputs
  • Controllable outputs: 2 relays, 2 TTL, 2 analog 10VDC max
  • View status and performance data on the Web with standard Web Browsers
  • Simple graphical, icon-based instrument setup with HyperWare II software
  • Easily created custom web pages via Drag n Drop graphics. No HTML programming required
  • FTP of logged data transferred via Browser, FTP client or Logger initiated
  • Email and SMS of alarms or status with data
  • Compact Flash memory expansion
  • Universal isolated analog inputs accept numerous sensor types
  • Expandable I/O (ILIM-7)
  • HyperWare II included for communications, programming, real-time viewing and file conversion, with full graphing capabilities