Lovibond 214050 IRim Infrared Data Transfer Module for Mc500 Colorimeters
Representative image only.
Lovibond Mfr # 214050 – Item # EW-99599-02
Optimize water analysis
  • For Lovibond water analysis equipment
  • 2 year warranty
PRODUCT OPTIONS
$383.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOrbeco-Hellige MC500 colorimeter
  • DescriptionIRim Infrared Data Transfer Module for Mc500 Colorimeters
  • Warranty2 year
Two year warranty
Key Features
  • For Lovibond water analysis equipment
  • 2 year warranty
More About this Item
Choose the following water analysis accessories to meet your application needs.
The IRiM uses state-of-the-art infrared technology to transmit data from the MC500 Multi-Parameter Colorimeter to one of 3 optional interfaces. Users can choose to connect to a Computer, a USB printer or a serial printer. The interface can be easily selected, and the choice is displayed by an LED function indicator. The user can switch between the interfaces using a “Select” button.
The IRiM comes with the software necessary to begin quickly and easily transfering data to your computer. As an option the data can be saved as an Excel spreadsheet or a .txt file. Measurement data can quickly be printed out, using a specified USB or serial printer connected to the IRiM.