Lovibond L147100 Test Kit , PH, 6.5 to 8.4
Representative image only.
Lovibond Mfr # 147100 – Item # EW-05542-45
Accurate and easy to use for routine water analysis
  • Continuous color disc
  • Tablet and powder reagent chemistries
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points6.5 to 8.4 pH
  • Tests Per Kit100
  • DescriptionTest Kit , PH, 6.5 to 8.4
  • Warranty1 year
1 Year Warranty
Key Features
  • Continuous color disc
  • Tablet and powder reagent chemistries
  • 1 year warranty
More About this Item
Simply add the tablet or powder pack reagent to the sample cell, allow it to dissolve per specific parameter instructions, rotate the color disc to match the color of the sample, and read off the concentration value.