Lovibond MD100 Colorimeter, Copper Kit

Representative image only.
Lovibond Mfr # 276085 – Item # EW-99561-85
Uses popular Powder Pack™, tablet, and liquid chemistries
  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant

PRODUCT OPTIONS

$502.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeLCD
  • Light SourceLED
  • Absorbance Accuracy±1 nm
  • Test ParametersCopper
  • Min Operating Temperature (° F)32
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)40
  • Number of Tests Included100
  • Data LoggingYes
  • Memory Size16 data sets
  • Battery1 x 9V battery (included)
  • Length (in)6
  • Width (in)3
  • Height (in)1 1/2
  • Range0.05 to 5.0 mg/L
  • DescriptionColorimeter, Copper Kit

Includes

three 24-mm sample cells with light-shield caps, reagents for 100 tests, foam-lined case, and four AAA batteries.

Key Features

  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant

More About this Item

The Lovibond MD100 series colorimeters are available in a wide variety of parameters using powder-pack chemistries. These rugged, waterproof meters feature a large backlit display, real-time clock, data logging capabilities, and a user calibration mode which allows you to calibrate against prepared standards.

(Note: Lovibond is the new name for Orbeco-Hellige--exactly the same product with a new name.)

Range for reagent: 0.05 to 5.0 mg/L

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat