Lovibond RP215-0 Powder Pillow, Sulfate 4 Reagent, 25 mL, 100 tests
Representative image only.
Lovibond Mfr # 532150 – Item # EW-99560-81
Premeasured reagents in convenient powder packs
  • Precise dosing with simple push-button operation
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersSulfate
  • Number of Tests Included100 x 25 mL
  • DescriptionPowder Pillow, Sulfate 4 Reagent, 25 mL, 100 tests
Key Features
  • Precise dosing with simple push-button operation
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.