Lovibond RS5250 Tube Test Set, Total and Acid Hydrolyzable Phosphorous, 50 tests
Representative image only.
Lovibond Mfr # 535250 – Item # EW-99560-80
Premeasured reagents in convenient powder packs
  • Precise dosing with simple push-button operation
PRODUCT OPTIONS
$93.50USD / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersTotal and acid-hydrolyzable phosphorous
  • Number of Tests Included50
  • DescriptionTube Test Set, Total and Acid Hydrolyzable Phosphorous, 50 tests
Key Features
  • Precise dosing with simple push-button operation
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.