Lovibond RS5300 Powder Pillow, Molybdate Reagent, High Range, 10 mL, 100 tests
Representative image only.
Lovibond Mfr # 535300 – Item # EW-99560-67
Premeasured reagents in convenient powder packs
  • Precise dosing with simple push-button operation
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersMolybdate, high range
  • Number of Tests Included100 x 10 mL
  • DescriptionPowder Pillow, Molybdate Reagent, High Range, 10 mL, 100 tests
Key Features
  • Precise dosing with simple push-button operation
More About this Item
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.