Low-Cost PVC Two-Way Ball Valve, 1/2" NPT(F) connectors
Representative image only.
Item # EW-07387-22
$24.15USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Cv8
  • Wetted MaterialsPVC
  • DescriptionLow-Cost PVC Two-Way Ball Valve, 1/2" NPT(F) connectors
More About this Item
Control flow with the 90°> rotating handle. Valves have seals made of Viton® fluoroelastomer and seats made of PTFE. Maximum pressure is 150 psi at 32°F
(10.3 bar at 0°C); maximum temperature is 140°F
(60°C at 1.7 bar).