Luer fittings, Polycarbonate fittings, male lock x male lock, rotating, Non-sterile
Representative image only.
Item # EW-06464-90
Easily transition to a luer fitting
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$9.10USD / Each
BUY MORE & SAVE
  • 1-9$9.10USD / Each
  • 10-49$8.19USD / Each
  • 50-99$7.74USD / Each
  • 100+$7.28USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 2Male luer lock
  • DescriptionLuer fittings, Polycarbonate fittings, male lock x male lock, rotating, Non-sterile
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available. Polycarbonate material has good chemical compatibility and can be sterilized via ethylene oxide or gamma irradiation.